VWR® Safety-Blue™, Hidrómetros con termómetros
Proveedor: VWR Collection
620-0924EA 107 EUR
620-0924 620-0925 620-0926 620-0928 620-0929
VWR® Safety-Blue™, Hidrómetros con termómetros
Hidrómetros
These safety sugar scale combined form thermo-hydrometers can determine the specific gravity, Brix and alcohol by volume.
  • Lead free ballast and glass with mercury-free blue spirit organic liquid fill
  • Individually serialised, safe and easy to clean up in the event of a breakage
  • Temperature standardisation of 20 °C with thermometer
Designed to measure sucrose in the food and beverage and other industries.
Certificaciones: ASTM
Información de suministro: Includes a multi-language statement of accuracy indicating traceability to NIST.
Precaución:More Product Information