VWR® TH300, Hygro/Temperature/Flow meter with Humidity / Temperature Probe
Proveedor: VWR Collection
620-1954EA 465 EUR
620-1954
VWR® TH300, Hygro/Temperature/Flow meter with Humidity / Temperature Probe
Higrómetros Termohigrómetros digitales
Digital precision handheld universal meter for humidity, temperature, dew point, distance, enthalpy and flow velocity, with additional thermocouple input in one. The plug-in probes are interchangeable without recalibration. The instrument is suitable for applications such as air conditioning, ambient air monitoring and flow measurement.
  • Double-display of humidity and temperature
  • Calculation of dew point temperature, dew point distance and enthalpy
  • Additional Nicr-Ni K-type socket for surface measurement
  • Maximum/minimum value memory, 'HOLD' function
  • Serial