VWR® Traceable®, USB Data Logging DUAL Thermo-Hygrometer, Excursion Trac
Proveedor: VWR Collection
620-2629EA 399 EUR
620-2629
VWR® Traceable®, USB Data Logging DUAL Thermo-Hygrometer, Excursion Trac
Higrómetros Termohigrómetros digitales
The VWR® Traceable® excursion trac USB data logging DUAL hygrometer with rolling memory structure monitors temperature and humidity readings in the laboratory with both internal and external sensors.
  • Memory capacity of 525 504 humidity and temperature observations
  • Four calibration points (25 °C and 50% RH for both internal and external sensors)
  • Temperature range: 0 to 65 °C (internal sensor), −20 to +70 ˚C (external sensor)
  • Humidity range: 0 to 95% non condensing (internal and external sensor)
  • LCD shows current temperature and humidity conditions, along