VWR® Traceable®, USB Data Logging Barometer
Proveedor: VWR Collection
620-2630EA 399 EUR
620-2630
VWR® Traceable®, USB Data Logging Barometer
Barómetros
The VWR® Traceable® excursion trac USB data logging barometer simultaneously monitors barometric pressure, temperature and humidity.
  • Memory capacity of 525000 temperature observations for barometric pressure, temperature and humidity
  • Calibration points: 25 °C for temperature, 50% RH for humidity, and 650, 850 and 1000 mbar for barometric pressure
  • Monitors barometric pressure, temperature and humidity readings with user-adjustable logging interval from 1 minute to 12 hours
  • This barometer has altitude compensation to convert local barometric pressure to sea