tempmate.®-S2 Single Use Temperature Data Logger, L1 15 °C, H1: 25 °C
Proveedor: tempmate
S2T-102
620-2833EA 587 EUR
620-2833
tempmate.®-S2 Single Use Temperature Data Logger, L1 15 °C, H1: 25 °C
Registradores de datos
Pre-programmed. Outstanding single-use temperature and humidity logger, for reliable cold chain monitoring. Integrated USB port and PDF and CSV generating software on-board.
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food grade packing
  • No additional hardware or software required
  • Detailed PDF and CSV report including graph and table
  • Intelligent LEDs for status monitoring
  • Extended shelf-life of two years
tempmate.®-S2 loggers are specially designed for temperature and/or humidity monitoring in any cold chain logistics.More Product Information