VWR® Traceable® Refrigerator/Freezer Digital Thermometers
Proveedor: VWR Collection
620-2838EA 68.68 EUR
620-2838 620-2839 620-2840
VWR® Traceable® Refrigerator/Freezer Digital Thermometers
Termómetros Termómetros digitales
Reliably monitor and be alerted to any issues affecting the contents of your fridge or freezer without even opening the door.
  • Alarm indicator lights up brightly to draw your attention to an immediate issue
  • Know about excursions before they happen with the pre-alert alarm light that glows orange when your fridge or freezer is within 1 degree of your alarm setting
  • Use the mute button to silence the buzzer alarm while you are investigating the issue
  • Staff onboarding is a breeze with an enhanced, more intuitive user experience including large buttons, high-
More Product Information
Limited Time Sale
Select items below are currently on sale