VWR®, Tiras indicadoras de temperatura
Proveedor: VWR Collection
620-9102EA 33.5 EUR
620-9102 620-9103 620-9104 620-9105 620-9106
VWR®, Tiras indicadoras de temperatura
Tiras de indicación de temperatura
These temperature indicating strips are designed for rapid measurements where conventional thermometers are impractical. They are suitable for use with equipment such as heating chambers and products which are sensitive at high temperatures.
  • Autoadhesivas
  • Respuesta en 1 segundo
  • Pueden almacenarse a 25 °C como máximo durante 2 años