Estereomicroscopios, Serie SFC-11
Proveedor: MOTIC
PX11426201
630-0694EA 450 EUR
630-0694
Estereomicroscopios, Serie SFC-11
Microscopios
The SFC-11 is the ideal stereomicroscope for both educational and industrial applications. Inclined at 45°, usage of this stereomicroscope is comfortable and fatigue free. Available in three turret magnification combinations [1×,2×; 1×,3×; and 2×,4×], thus providing a choice on magnification needed for the application. The standard working distance is sufficient for majority of applications without the need for addition of auxiliary objectives.
  • Objective turret magnification changer (2×, 4×)
  • Standard working distance of 95 mm
With a variety ofMore Product Information