Ring light, VisiLED Darkfield
Proveedor: SCHOTT AG Lighting and imaging
400300
630-1196EA 899 EUR
630-1196
Ring light, VisiLED Darkfield
Luces Anillo de luces
VisiLED Darkfield ring light provides intensive illumination in the incident darkfield, enhancing contrasts of flat structured surfaces.
  • Easily adaptable to microscope objectives by means of thumb screw and adaptor rings
  • Homogenous illumination
  • Best white quality: Typically 6000 K
  • Long operating life: Typically 50000 hours by actively controlling the LED chip temperature
  • Segments controllable in different preset modes for optimum contrasting