Benchtop cryostat
Proveedor: Slee
10140000 10142000
631-0558EA 13200 EUR
631-0558 631-0560
Benchtop cryostat
Microtomas Micrótomos
The compact design makes this instrument a nice backup for installed high-end cryostats.
  • Integrated heat extractor with intuitive control panel
  • Automatic defrost plus UV-C disinfection
  • Viewing window made of frost-free dual insulation glass
  • Height adjustable support stand and orientation of specimen holder in three planes
Designed for histology and pathology, research laboratories with special attention to hygiene and ease of handling.
Cutting stroke: 52 mm
Max specimen size: 25 mm
Información de suministro: Includes an operation manual, twoMore Product Information