Handheld refractometer, HHR-2N
Proveedor: Atago
2522
635-0140EA 376 EUR
635-0140
Handheld refractometer, HHR-2N
Refractómetros Refractómetros de mano
This refractometer is designed to determine the water content in honey.
  • Scale to measure the percentage of water
  • Refractive index (RI) of sucrose
  • Determination of moisture in honey
Specifically suitable for use in food applications.