Immersion flow cups, ISO 2431
Proveedor: Industrial Physics
VF2093 VF2092 VF2091 VF2090 VF2224 VF2222 VF2185
649-0010EA 174.96 EUR
649-0010 649-0009 649-0008 649-0007 649-0014 649-0013 649-0011
Immersion flow cups, ISO 2431
Viscosímetros Copas de flujo
Titanium anodized aluminum or stainless steel cups designed with fixed stainless steel nozzle and handle, for measuring coatings and other fluids.
Certificaciones: ISO 2431