Supelco® Discovery® BIO C5 Reversed-Phase Solution to Protein and Peptide Separation Challenges
Proveedor: Merck
567226-U 567227-U 567228-U 567229-U 567230-U 567231-U 567232-U 567234-U 567278-U 567280-U 567281-U 65511-U 568402-U 568420-U 568421-U 568422-U 568423-U 568430-U 568472-U
567226-UEA 677 EUR
567226-U 567227-U 567228-U 567229-U 567230-U 567231-U 567232-U 567234-U 567278-U 567280-U 567281-U 65511-U 568402-U 568420-U 568421-U 568422-U 568423-U 568430-U 568472-U
Supelco® Discovery® BIO C5 Reversed-Phase Solution to Protein and Peptide Separation Challenges
Columnas de cromatografía
Discovery® BIO Wide Pore reversed-phase columns satisfy the need for efficiency, selectivity, scalability, and reproducibility for RP HPLC analyses of proteins, peptides, and small biomolecules.
  • Enhanced stability and lifetime compared to conventional C4 phases
  • Better protein and peptide resolution compared to leading RP-HPLC FPP columns
  • High efficiency for peptide mapping
  • Low-bleed, inert surface chemistry make them ideal for LC-MS applications
  • Reliable reproducibility run -to-run, column-to-column, batch-to-batch
  • Separation are completely scalable from