Titrators, Karl Fischer, volumetric, TitroLine® 7500 KF
Proveedor: SI Analytics
285220810 285220820 285220830
661-0138EA 7210 EUR
661-0138 661-0139 661-0140
Titrators, Karl Fischer, volumetric, TitroLine® 7500 KF
Valoradores
TitroLine® 7500 KF is a volumetric generalist for a wide range of uses. Applications include KF volumetry, dead-stop titrations (SO₂, bromine number), dosing tasks and preparation of solutions. Ideal for KF analysis in pharmaceutical, chemical, food and mineral oil industries.
  • Fast, easy and precise
  • Ready to use methods for different applications (e.g. titre determination, blank value)
  • High visibility, full colour display with online titration curve, can be easily viewed from a distance and extreme angles
  • Storage of results via USB port (PDF and