Sera-Mag Streptavidin-Coated Magnetic Beads and SpeedBeads
Proveedor: Cytiva
66152104010150 66152104010350 66152104011150 30152103010150 30152103010350 30152103011150 30152104010150 30152104010350 30152104011150 30152105010150 30152105010350 30152105011150
66152104010150.EA 935 EUR
66152104010150. 66152104010350. 66152104011150. 30152103010150. 30152103010350. 30152103011150. 30152104010150. 30152104010350. 30152104011150. 30152105010150. 30152105010350. 30152105011150.
Sera-Mag Streptavidin-Coated Magnetic Beads and SpeedBeads
Perlas Bolas magnéticas
Sera-Mag Streptavidin-Coated Magnetic Beads and SpeedBeads exhibit high affinity and sensitivity for biotinylated target molecules, along with fast reaction kinetics, increasing throughput and precision for genomic and proteomic applications.
  • Available with low (2500 to 3500 pmol/mg), medium (3500 to 4500 pmol/mg) or high (4500 to 5500 pmol/mg) nominal biotin binding capacities for optimising assay development
  • Low dissociation constant to tightly bound ligands
  • Low non-specific binding for greater precision and accuracy
  • Available in Sera-Mag SpeedBeads and