Conductivity meters, handheld, Seven2Go™ S3 / S7 Pro
Proveedor: Mettler - Toledo
30207873 30207956 30207963 30207962 30207961 30207957 30207955 30207954
662-1843EA 1720 EUR
662-1843 662-1841 662-1855 662-1854 662-1853 662-1842 662-1840 662-1839
Conductivity meters, handheld, Seven2Go™ S3 / S7 Pro
Medidores electroquímicos Medidores de conductividad
These robust meters provide durability and one-handed operation, making them ideally suited for mobile applications in the laboratory, at-line or outdoors. Thanks to their IP 67 protection and drop test resistance, the devices are able to withstand harsh and demanding environments.
  • New intuitive menu guarantees ease of use
  • T-Pad with smart button arrangement enables easy and fast navigation
  • Instrument automatically recognises InLab ISM sensors and stores sensor ID, serial number, and last calibration data
  • Linear temperature compensation
Only model S7More Product Information