Special pH electrodes, IDS (Intelligent Digital Sensors), SenTix®
Proveedor: WTW
103743 103751 103752 103753
662-1874EA 356 EUR
662-1874 662-1875 662-1876 662-1877
Special pH electrodes, IDS (Intelligent Digital Sensors), SenTix®
Electrodos Electrodos pH
These special pH electrodes extend the range of the IDS series for enhanced applications in Life Sciences and related applications.
  • Intelligent: Electrodes are automatically recognized by the meter, each sensor is exactly identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • Measurement integrity benchmarking of the cells by QSC (Quality Sensor Control)
  • IDS electrodes store their serial number and their calibration records
  • Easy exchange of electrodes thanks to calibration data stored in the sensor
  • GLP support due to