BOD and respirometric measurements, Bluetooth®, OxiTop®-IDS
Proveedor: WTW
209700 209701 209702 209704 209732 2FD56W 2FD57W 209714 209716 209711 209713 209721 209723 209725 209727 209729 209731
WTWA209700EA 553 EUR
WTWA209700 WTWA209701 WTWA209702 662-2317 WTWA209732 662-2318 662-2319 705-1185 705-1186 705-1181 705-1184 705-1182 705-1187 662-2345 705-1188 705-1189 705-1183
BOD and respirometric measurements, Bluetooth®, OxiTop®-IDS
DBO, Demanda biológica de oxígeno
OxiTop®-IDS (Intelligent Digital Sensors) is a modern measuring instrument for use in all kinds of respirometric studies. The versatile OxiTop®-IDS is suitable for both aerobic or anaerobic examinations. All heads can be used independently from any meter for normal BOD measurements between one and seven days.
  • Representation of curves for monitoring
  • Parallel measurement of different examinations and methods
  • Backlit graphic LCD supports parameter input and user information also polling of interim results
  • Direct calculation no need to convert data - direct