pH meter, SevenDirect SD20
Proveedor: Mettler - Toledo
30671550 30671554 30671555 30671556 30671557 30671558 30671559 30671560 30671561
662-2390EA 1330 EUR
662-2390 662-2394 662-2395 662-2396 662-2397 662-2398 662-2399 662-2400 662-2402
pH meter, SevenDirect SD20
Medidores electroquímicos Medidores de pH
Robust bench top meter for accurate pH determinations in the lab and near production.
  • Large colour touch screen provides direct measurement and easy data entry
  • Instructions guide users through the sensor calibration process
  • Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)
The large colour touch screen offers a direct menu access, fast dataMore Product Information