pH half cells, InLab®
Proveedor: Mettler - Toledo
51343195 51343196
662-2874EA 198 EUR
662-2874 662-2875
pH half cells, InLab®
Electrodos Electrodos pH
pH electrode with glass body and S7 screw head. Recommended for applications where the life time of the pH electrode is significantly shorter than the reference electrode.
InLab® Mono
- Extended reference lifetime
- Glass body gives protection against corrosive and organic samples
- Detachable cable: Different cables can be connected to the S7 screw head allowing use with a wide range of meters
InLab® Mono Plus
- Extra strong glass membrane
- Maximum temperature 130 °C
- Extended reference lifetime
- Glass body gives protection against corrosive and organic samples
- Detachable cable:More Product Information