Electrodos de conductividad
Proveedor: Mettler - Toledo
51302255 51302256 51344020 51344022 51344024 51344120 51344122 51344124 51344126 51344128 51344030 51344031 30014160 51340336 51344026
662-2814EA 550 EUR
662-2814 663-0061 663-0148 663-1001 663-0149 663-0150 663-0271 663-0272 663-0273 663-0274 663-0231 663-0232 663-0266 663-0267 663-0310
Electrodos de conductividad
Electrodos Sensores de conductividad
These conductivity probes with integrated temperature sensor (NTC 30 kΩ) are suitable for different sample types. They all provide automatic temperature compensation.
Suitable for:
InLab 710/InLab 720/InLab 725: SevenEasy™, SevenMulti™, FiveEasy™, FiveGo™
InLab 731/InLab 741: SevenExcellence™, SevenCompact™
InLab 737/InLab 738: SevenGo Duo™ SG23, SevenGo Duo pro™ SG78
InLab 742: SevenGo™ SG3, SevenGo pro™ SG7