pH/mV/°C/ISE meter, bench, Lab 845
Proveedor: SI Analytics
285206470
663-0323EA 572 EUR
663-0323
pH/mV/°C/ISE meter, bench, Lab 845
Medidores electroquímicos Medidores de pH
This small benchtop meter with intuitive operation and robust aluminum housing is perfect for training and routine purposes.
  • User-friendly design, numerical access protection possible
  • Graphic backlight LCD display
  • Data logger (4000 data sets)
  • Min/Max values with date and time stamp
  • Aluminum desk housing (IP 40/DIN EN 60529)
  • Developed and made in Germany
Información de suministro: Meter only is supplied with power supply.
Lab 845 Set/BL19pH also includes stand, pH electrode BlueLine 19 pH, and DIN buffers in ampules (6 pieces).
Lab 845 Set/BL25pH alsoMore Product Information