Conductivity meter, bench, 703
Proveedor: KNICK
703
663-2101EA 2630 EUR
663-2101
Conductivity meter, bench, 703
Medidores electroquímicos Medidores de conductividad
This bench top meter can be operated with either 2-electrode or 4-electrode sensors. In conjunction with the Knick ZU 6985 4-electrode sensor, the meter functions reliably over a wide conductivity range from <1,00 to >1000 mS/cm.
  • Calibrated analog recorder output, galvanically isolated
  • Sensoface® function monitors the conductivity sensor and measuring equipment and provides information on sensor selection and handling
  • Automatic calibration with standard solutions
  • RS-232 interface for computer and printer
  • Simultaneous display of two measurement values;