Hexyl chloroformate ≥97.0% (por GC), LiChropur™ for GC derivatisation, Supelco®

Proveedor: Merck

67389-10ML
67389-10MLEA 227 EUR
67389-10ML
Hexyl chloroformate ≥97.0% (por GC), LiChropur™ for GC derivatisation, Supelco®
Hexyl chloroformate
Formula: C₇H₁₃ClO₂
Número MDL: MFCD00000651
Núm. CAS: 6092-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b79ec934582a079444d9630866c168eac2971e01&vid=b7d9ee32685639c74411ca16981d42213037ff17&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR