Potasio cloruro 99,0-100,5, granular, AR® ACS, Macron Fine Chemicals™

Proveedor: Avantor Performance Materials
Atención

Sinónimos: Potassiumchloride

6858-26EA 3419.44 EUR
6858-26 6858-20 6858-04
Potasio cloruro 99,0-100,5, granular, AR® ACS, Macron Fine Chemicals™
Potasio cloruro
Formula: KCl
MW: 74.55 g/mol
Pto. Ebullición: 1420
Pto. de fusión: 773
Densidad: 1.98 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011360
Núm. CAS: 7447-40-7
EINECS: 231-211-8
Índice Merck: 14,07621

Order Now

Resultados test especificaciones

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (as KCl) (dried basis) 99.0 - 100.5 %
ACS - Barium (Ba) Passes Test
ACS - Bromide (Br) ≤0.01 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Chlorate and Nitrate (as NO₃) ≤0.003 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iodide (I) ≤0.002 %
ACS - Iron (Fe) ≤3 ppm
ACS - Magnesium (Mg) ≤0.001 %
ACS - pH of 5% Solution at 25°C 5.4 - 8.6
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Sodium (Na) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.001 %
ACS - Loss on Drying at 105°C ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR