Organic reference standard, Dimethyl disulfide, analytical standard, Supelco®

Proveedor: Merck

68986-1ML
68986-1MLEA 50.6 EUR
68986-1ML
Organic reference standard, Dimethyl disulfide, analytical standard, Supelco®
Normativas Estándares orgánicos
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c537a2f2e8bf4427f675fe44267be613a0fe4ebf&vid=c537a2f2e8bf4427f675fe44267be613a0fe4ebf&prid=web&clid=SAPDAM

Order Now

ESPECIFICACIONES

Descripción Dimethyl disulfide
Environmentally Preferable
CAS No. 624-92-0
Formula C₂H₆S₂
Melting point –85 °C (lit.)
Punto de ebullición 109 °C (lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR