Anti-IL-10 Rat Monoclonal Antibody [clone: JES5-2A5]

Proveedor: Tonbo Biosciences
70-7102-U100
70-7102-U100EA 101 EUR
70-7102-U100
Anti-IL-10 Rat Monoclonal Antibody [clone: JES5-2A5]
Anticuerpos
The JES5-2A5 antibody is specific for mouse IL-10. IL-10 has numerous biological activities including the ability to inhibit both the synthesis of Th1 cytokines and proliferation of T cells. In the mouse, IL-10 is produced by phagocytic cells, conventional dendritic, cells, T cells, B cells, and NK cells. The JES5-2A5 antibody is a neutralizing antibody.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: JES5-2A5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR