Dual-gel electrophoresis system, Owl™ P8DS
Proveedor: THERMO OWL SCIENTIFIC
P8DS-1 P8DS-2 P8DS-3
730-0315EA 2220 EUR
730-0315 700-2340 700-2341
Dual-gel electrophoresis system, Owl™ P8DS
Sistemas de electroforesis Sistemas verticales de electroforesis en gel
The Owl™ P9DS dual gel system is designed for quick and easy casting and running of one or two gels on the same device. When used with the available combs, spacers, alumina plates and glass, this system is an ideal tool for second dimension protocols of 2-D electrophoresis, SDS and agarose gels, and native PAGE electrophoresis.
  • Agarose plug method makes gel casting simple and quick
  • Gel assemblies do not have to be moved once set up
  • Two individual steps of casting and running are combined
  • Simple clamping mechanism makes loading glass cassettes, and most