Soportes para microtubos, reversibles, con tapa
Proveedor: Thermo Fisher Scientific
8660 8670 8680 8601
700-2824EA 0 EUR
700-2824 700-2825 700-2826 MLBP8601
Soportes para microtubos, reversibles, con tapa
Gradillas Tube Racks
Made of PP.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Sturdy
These racks are for microcentrifuge tubes with molded alphanumeric grid and also assorted colours are available.