Flatpack chambers
Proveedor: BTX
45-0109
700-4813EA 653 EUR
700-4813
Flatpack chambers
Reactivos de ácidos nucleicos Reactivos para transformación
Flatpack chambers are primarily used for prokaryotic applications (bacterial or yeast transformations), however they are also often used for high efficiency stem cell transfection.
  • The flow-through construction of the 0,56 mm gap has a volume capacity from 10 to 85 μl. This design provides the unique combination of small sample volumes with field strengths as high as 40 kV/cm.
The flatpack chamber with 1,83 mm gap has a three-ply solid sandwich construction of stainless steel and mylar plastic and holds a volume of 1,5 ml, ideal for stems cells.
Flatpack chambers are gamma sterilised in individual packages.
Información de suministro: Supplied in sets of 50 and may be used in the BTX safety stand (available separately, please enquire).