Test strips, ascorbic acid, QUANTOFIX®
Proveedor: MACHEREY-NAGEL
91314
705-0315EA 36.8 EUR
705-0315
Test strips, ascorbic acid, QUANTOFIX®
Kits para test medioambiente
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are for the rapid and reliable determination of Ascorbic acid/vitamin C in food. The test strips' easy dip-and-read-procedure provides reliable results within 30 seconds.
Ascorbic acid or vitamin C is naturally found in many foods and vegetables. It is also added to juices or fruits as stabilizing and reducing agent. Thus, ascorbic acid and vitamin C are important parameters in the fruit producing and food processing industry. QUANTOFIX® ascorbic acid test strips allow the quick and easy determination of vitamin C in fruit juices as well as on fresh cut surfaces of fruits and vegetables.
Gradation: 0 · 50 · 100 · 200 · 300 · 500 · 700 · 1000 · 2000 mg/L vitamin C
Colour: yellow → green-blue