Accessories for comparators
Proveedor: Lovibond
221080 221130 230020 230030 230060 230070 230110 230170 232010 234010 234060 234081 234090 235800 236000 238500 283110 283120 283130 283450 283580 283630 283720 284120 284150 284160 284170 292260 234020 284130 242600 242900 221010 230320 230330 232910 234110 283410 283510 283520 283610 283700 283730 283920 283140 230180 235310 235320 283470 230050 230040 283460 233920 232070 232090 230290 230190 230100 283680 283960 236700 230650 230440 230270 230310 221050 221190 230340 230420 231300 230210 230470 230620 283950 234140 283440 235900 221140 233930 237000 221100 221110 230205 297970 242801 295920
705-0614EA 215 EUR
705-0614 705-0616 705-0619 705-0620 633-0167 633-0168 705-0622 705-0623 705-1041 633-0179 633-0181 633-0182 633-0183 705-0567 705-0637 705-0668 633-0188 633-0189 633-0190 633-0193 705-0640 705-0641 705-0643 633-0197 633-0198 633-0199 633-0200 705-1055 633-0180 TINT284130 705-1060 705-1061 705-0612 705-0627 705-0628 705-0636 705-0903 705-1043 705-1044 705-1045 705-1047 705-1049 705-1050 705-1053 633-0191 633-0169 633-0185 633-0186 633-0195 633-0166 633-0165 633-0194 633-0177 633-0175 633-0176 633-0172 633-0170 705-0621 705-0642 705-0644 705-0638 705-0634 705-0632 705-0625 705-0626 705-0613 705-0618 705-0629 705-0631 705-0635 705-0624 705-0633 705-1039 705-1054 633-0184 633-0192 633-0187 705-0617 633-0178 705-0639 705-0615 705-0648 633-0171 633-0201 TINT242801 705-1056
Accessories for comparators
Colorímetros Kits de comparación
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Información para pedidos: The most widely used discsMore Product Information