Copan Transystem® Traditional Swab Collection and Transport System for Aerobic and Anaerobic Bacteria
Proveedor: Copan
124C.USE 144C 111C.USE 113C.USE 117C.USE 132C.USE 147C 148C 145C 134C.USE 139C.USE 143C.USE 125C.USE 136C.USE 126C.USE 138C.USE 414C
COPA124C.USEEA 2250 EUR
COPA124C.USE 710-0450 710-0443 710-0445 710-0446 710-0449 710-0453 710-0454 710-0451 710-0924 710-0926 710-0927 710-0923 710-0925 COPA126C.USE COPA138C.USE COPA414C
Copan Transystem® Traditional Swab Collection and Transport System for Aerobic and Anaerobic Bacteria
Hisopos Hisopos para transporte
Our Transystem™ family comprises different media for the efficient and safe transport of many bacterial strains. Choose between liquid or solid Amies and Stuart medium for aerobic culture, rapid antigen, and molecular testing; opt for gel Cary-Blair medium - with or without charcoal - for aerobic and anaerobic cultures. Bacterial transport basics. A simple, low bioburden, device for every investigation. Just uncap, collect and recap the tube.
  • Swab variety: Various swab sizes to reach different collection sites
  • Media assortment: Choice of liquid or solid