Placa de Petri, Nalgene®
Proveedor: Thermo Fisher Scientific
5500-0010
710-4105EA 63 EUR
710-4105
Placa de Petri, Nalgene®
Placas de cultivo Placas de Petri
Polymethylpentene (PMP). Useful for biotechnology applications as resistant to cell adherence.
  • Shatterproof
  • Transparent
  • Autoclavable
  • Chemically resistant
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact
Certificación ambiental NAR Certificate of Retirement