Anti-IgG Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Tonbo Biosciences
72-8078-M001
72-8078-M001EA 147 EUR
72-8078-M001
Anti-IgG Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Sheep anti-Rabbit IgG (H&L) - Affinity Pure, HRP Conjugate

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Sheep
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR