VWR® Q Path® MicroStar V, Casetes, en racks
Proveedor: VWR Collection
720-2091EA 374 EUR
720-2091 720-2092 720-2093 720-2094 720-2095 720-2096 720-2097 720-2098 720-2090
VWR® Q Path® MicroStar V, Casetes, en racks
Casetes para histología Casetes de procesamiento de biopsias
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with welded disposable lid, supplied in racks for Leica and Sakura cassette printers. 2000 cassettes packed 40/rack.
  • Easy to use cassette with small opening
  • Writing at the front and on both sides
  • Cassettes with ridges, which makes it easier to be opened after impregnation