CellStor Pot™ specimen container
Proveedor: CELLPATH
BAF-9000-08A BAF-9100-08A BAF-6000-08A
720-2324EA 26 EUR
720-2324 720-2325 CLLPBAF-6000-08A
CellStor Pot™ specimen container
Recipientes de muestras Contenedores, uso general
Prefilled specimen containers.
  • Leak-proof lid for safe transportation and specimen storage
  • With traceable badge number, safety data sheet in the packaging