Fine dosage syringes, Omnican®-F
Proveedor: B. Braun
9161502S
720-2560EA 63.2 EUR
720-2560
Fine dosage syringes, Omnican®-F
Jeringas Jeringas médicas
PP/PS fine dosage syringes, 1 ml, ideal for tuberculin, heparin and allergy testing. Easy aspiration up to the maximum volume. The integrated needle (Ø 0,30 mm, length 12 mm, 30 G) ensures no dead space and no wasted medication. PVC-free.
  • Good contrast, permanent black markings, with intervals of 0,01 ml
  • Plunger gasket, made of synthetic rubber - extremely slow aspiration or injection, smoother plunger action with double sealing ring
  • Safe plunger backstop
  • Transparent protective cap for needle
Envase: Dispenser box of 100 units.