Thermal cyclers, Mastercycler® nexus
Proveedor: EPPENDORF
6331000.041 6333000.049
731-0429EA 0 EUR
731-0429 731-0675
Thermal cyclers, Mastercycler® nexus
Termocicladores PCR
The Mastercycler® nexus brings the reliability and ease of use of an Eppendorf® cycler at a good price. Low energy consumption, easy extension to 3 units and a small footprint make the Mastercycler® nexus both cost effective and an efficient use of laboratory space. It can handle all kinds of PCR consumables, from low volume up to 0,5 ml PCR tubes. All common PCR plates fit and, if throughput becomes higher, it is possible to connect an additional one or two Mastercycler nexus eco units.
  • Flexlid™ concept allows use of different consumables with automatic