Adhesive sealing film for microplates, QuickSeal Gas Perm™
Proveedor: iST Scientific Ltd
IST-124-080SS IST-124-080LR IST-124-080LS IST-124-080SR IST-124-080TS
731-0622EA 248 EUR
731-0622 731-0619 731-0621 731-0620 731-0623
Adhesive sealing film for microplates, QuickSeal Gas Perm™
Juntas Sellos de microplaca
Transparent perforated gas permeable film. Suitable for bacterial culture, eukaryotic cell culture.
  • Peelable and pierceable
  • Seal integrity range: −20 to +80 °C