VWR®, Deep Well Plates, Square Well
Proveedor: VWR Collection
732-3323EA 247 EUR
732-3323 732-3325 732-3795 732-3796 732-3797 732-3806 732-3810 732-3811 732-3866
VWR®, Deep Well Plates, Square Well
Microplacas Placas Deep Well
These deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
  • Chemical resistant
  • Free from DNase and RNase
  • V-bottom well shape maximises liquid removal and aids resuspension
  • U bottom suited for washing or stirring samples
  • No inner edges to interfere with magnetic bead collection
  • Raised well rims to improve heat sealing
  • Conical base aids sample concentration, reconstitution and centrifugation
Minimum order quantity of 40More Product Information