96-well solid black and white polystyrene microplates, Corning®
Proveedor: Corning
3362 3600 3650 3688 3693 3694 3792 3912 3915 3916 3917 3922 3925 3605 3686 3642 3990 3993 3991 3875 4591 3789A
734-1549EA 602.96 EUR
734-1549 732-5515 732-5522 734-1623 734-1625 734-1626 734-1643 734-1662 734-1663 734-1664 734-1665 734-1666 734-1667 732-5517 734-1622 734-4117 732-5596 734-4171 734-4169 734-4153 CORN4591 CORN3789A
96-well solid black and white polystyrene microplates, Corning®
Microplacas Placas de cultivo celular
These 96-well solid black and white polystyrene microplates are designed to reduce well-to-well cross-talk.
White microplates enhance luminescent signals and have low background luminescence.
Black microplates have low background fluorescence and minimise light scattering.
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact
Certificación ambiental RECs, ISO 14001 - Kennebunk