Verso™ qRT-PCR Kits
Proveedor: Thermo Fisher Scientific
AB-4100/A AB-4100/C AB-4101/A AB-4101/C AB-4102/A AB-4102/C AB-4104/A AB-4104/C AB-4105/A AB-4105/C AB-4106/A AB-4106/C
733-1112EA 864 EUR
733-1112 733-1114 733-1117 733-1119 733-1122 733-1124 733-1127 733-1129 733-1132 733-1134 733-1137 733-1139
Verso™ qRT-PCR Kits
Reactivos de ácidos nucleicos qPCR/RT-qPCR Enzymes and Kits
The Verso™ qRT-PCR system combines the Verso™ RT enzyme with ABsolute™ Blue qPCR master mix to provide maximum convenience and versatility. Available in a 1- or 2-step format, as well as multiple pack sizes, to suit all application and throughput requirements.
  • The combining of Verso™ enzyme and Thermo-Start™ Taq polymerase gives sensitive results from low copy number RNA template
  • Inert blue dye for easy visualisation of the qRT-PCR reaction
  • No need to perform an RNase H digestion after cDNA synthesis, therefore streamlining the reaction protocol
  • RT enhancer saves time by eliminating the need for DNase treatment of samples
  • Multiple priming options for optimisation of specific RNA templates
Order Now
ESPECIFICACIONES
Environmentally Preferable
Aplicación RT-qPCR dye
One step Yes