FlashGel™ DNA system
Proveedor: LONZA
57023 57026 57032 57029 57031 57067 57051
733-1611EA 360 EUR
733-1611 733-1610 733-1615 733-1613 733-1614 LONZ57067 LONZ57051
FlashGel™ DNA system
Sistemas de electroforesis Sistemas horizontales de electroforesis en gel
FlashGel™ DNA system offers a rapid method for separating DNA and watching DNA migration as it happens. The system consists of enclosed, disposable, precast agarose FlashGe™ DNA cassettes and an electrophoresis unit with built in transilluminator (FlashGel™ dock) that provides both separation and detection.
  • Separates DNA fragments 10 bp to 4 kb in 2 to 7 minutes
  • Gel runs can be monitored at the bench, in real time without UV light
  • At least five times more sensitive than gels stained with ethidium bromide
Certificaciones: Some components and technologyMore Product Information