Cell culture dishes, poly-D-lysine, Corning® BioCoat™
Proveedor: Corning
354550
734-0201EA 138 EUR
734-0201
Cell culture dishes, poly-D-lysine, Corning® BioCoat™
Placas de cultivo Placas para cultivo celular
TC-treated gridded culture dishes.
Corning® BioCoat™ products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.