Placas multipocillos con poli-L-lisina y laminina Corning® BioCoat™
Proveedor: Corning
354595 354596 354619
734-0229EA 308 EUR
734-0229 734-0230 734-0236
Placas multipocillos con poli-L-lisina y laminina Corning® BioCoat™
Microplacas Placas de cultivo celular
PS, coated with mouse laminin. Laminin, a major structural component of basement membranes, has many varied functions that are mediated by binding to various components of the basement membrane (for example, collagen IV) and to cell-surface receptors. Laminin-coated cell culture plates have a number of applications, including the promotion of cell adhesion, proliferation and differentiation of a variety of cell types, particularly neurons, epithelial cells, myocytes and myoblasts.
Laminin source: Engelbreth-Holm-Swarm (EHS) mouse tumor. Tested for ability to initiate differentiation (More Product Information