Extracto de hipófisis bovina (BPE), Corning®
Proveedor: Corning
354123
734-1311EA 171 EUR
734-1311
Extracto de hipófisis bovina (BPE), Corning®
Suplementos y aditivos de cultivo celular
BPE es un suplemento ampliamente utilizado para cultivar una variedad de células epiteliales y endoteliales.
BPE contains growth factors (e.g., basic FGF) and hormones.
Tested for ability to promote proliferation of Fetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.
BPE contains growth factors (e.g., basic FGF) and hormones.