96-well plates, polypropylene, without lids
Proveedor: Corning
3355 3356 3357 3359 3363 3364 3365 3343 3344
734-1781EA 403.56 EUR
734-1781 734-1782 734-1545 734-1547 734-1550 734-1551 734-1552 732-0484 732-0485
96-well plates, polypropylene, without lids
Microplacas Microplacas HTS
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or non sterile
Información para pedidos: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.