Centrifuge tubes, sterile, polypropylene, Corning®
Proveedor: Corning
431123 430776
734-1880EA 296.97 EUR
734-1880 734-1863
Centrifuge tubes, sterile, polypropylene, Corning®
Tubos Tubos de centrífuga
PP, with HDPE plug seal cap, sterile. Polyetherimide cushions.
  • Moulded graduations, conical bottom
  • Tubes are sterile and certified non pyrogenic
  • Support cushions must be used unless the rotor has appropriately shaped V-bottom holders
Ideal for applications requiring large volume centrifugation.
They are perfect for large volume centrifugation applications. They incorporate plug seal caps and have molded graduations.
Información para pedidos: Non sterile cushion sold separately.
Envase: A granel, 6/bolsa.