Cell culture vessels, Corning® HYPERStack®
Proveedor: Corning
20012 20013 20036 20037
734-3377EA 3000 EUR
734-3377 734-3378 734-3379 734-3380
Cell culture vessels, Corning® HYPERStack®
Matraces Matraces de cultivo de células
The Corning® HYPERStack® cell culture vessel is a closed system that allows fluid manipulations to occur through preassembled tubing sets. Two ³/₈" ID tubing ports for filling and venting allow filling via bag assembly, sterile bottle connection or pumping. The attached vent filters are 0,2 μm pore, hydrophobic membranes that allow air displacement during filling while minimising the risk of contamination.
  • Features 18,000 cm² cell growth area
  • Closed system approach with integrally incorporated USP
  • Class VI certified C-FLEX tubing for liquid handling